CAS RN 3056570-19-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ AZ' '9567
Potent MAT2a (methionine adenosyltransferase 2a) inhibitor