accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3058-01-3

IUPAC Name: 3-methylhexanedioic acid
Synonyms: 3-methyladipic acid
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: SYEOWUNSTUDKGM-UHFFFAOYNA-N
SMILES: CC(CCC(O)=O)CC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results