Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyladipic Acid 99.0+%, TCI America™
SDP

Catalog No. M071925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M071925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M071925G Supplier TCI America Supplier No. M071925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3058-01-3
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00002804
InChI Key SYEOWUNSTUDKGM-UHFFFAOYNA-N
Synonym 3-methyladipic acid, hexanedioic acid, 3-methyl, +-3-methyladipic acid, beta-methyl-adipic acid, 3-methyl-hexanedioic acid, 3-methyl adipic acid, 3-methyl-adipic acid, acmc-20ah5v, acmc-1cr8a, hexanedioic acid,3-methyl
PubChem CID 12292
ChEBI CHEBI:68503
IUPAC Name 3-methylhexanedioic acid
SMILES CC(CCC(=O)O)CC(=O)O

Specifications

Melting Point 100°C
Color White-Yellow
Boiling Point 205°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyladipic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.