accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOOONa

CAS RN 305808-14-4

IUPAC Name: sodium 2-formylbenzene-1-sulfonate
Synonyms: sodium 2-formylbenzenesulfonate
Molecular Weight (g/mol): 208.16
Molecular Formula: C7H5NaO4S
InChi Key: ADPUQRRLAAPXGT-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O
Molecular Weight (g/mol) 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results