Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Formylbenzenesulfonic acid sodium salt hydrate

Catalog No. 239380100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-938-0100G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-938-0100G Supplier Sigma Aldrich Supplier No. 239380100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Formylbenzenesulfonic acid sodium salt hydrate

Specifications

CAS 305808-14-4
Molecular Formula C7H5NaO4S -+ xH2O
Linear Formula HCOC6H4SO3Na -+ xH2O
Quantity 250 mg
Synonym Benzaldehyde-2-sulfonic acid sodium salt
Molecular Weight (g/mol) 208.17 (anhydrous basis)
Percent Purity 75%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.