accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 306-03-6

IUPAC Name: hydrogen 8-methyl-8-azabicyclo[3.2.1]octan-3-yl (2S)-3-hydroxy-2-phenylpropanoate bromide
Synonyms: hydrogen hyoscyamine bromide
Molecular Weight (g/mol): 370.29
Molecular Formula: C17H24BrNO3
InChi Key: VZDNSFSBCMCXSK-ZQXFGJFASA-N
SMILES: [H+].[Br-].CN1C2CCC1CC(C2)OC(=O)[C@H](CO)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results