Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyoscyamine HBr, MP Biomedicals

Catalog No. ICN207727 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN207727 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN207727 Supplier MP Biomedicals, Inc Supplier No. 0520772710

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 306-03-6
Molecular Formula C17H24BrNO3
Molecular Weight (g/mol) 370.287
InChI Key VZDNSFSBCMCXSK-BXSBZORQSA-N
Synonym hyoscyamine hydrobromide, daturine hydrobromide, unii-iwt50p9s79, l-hyoscyamine hydrobromide, hyoscyamine hydrobromide usp, --atropine hydrobromide, c17h23no3.hbr, 1alphah,5alphah-tropan-3alpha-ol--tropate ester hydrobromide, unii-6vkw7r97zu component vzdnsfsbcmcxsk-pgqienjjsa-n
PubChem CID 9929278
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2S)-3-hydroxy-2-phenylpropanoate;hydrobromide
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.Br

Specifications

Quantity 10 g
Formula Weight 370.28
Chemical Name or Material Hyoscyamine HBr
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.