accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3060-46-6

OHOHN(S)CH3CH3H3C

CAS RN 3060-46-6

IUPAC Name: (2S)-4-methyl-2-(methylamino)pentanoic acid
Synonyms: L-leucine, N-methyl-
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: XJODGRWDFZVTKW-LURJTMIESA-N
SMILES: CN[C@@H](CC(C)C)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)