accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30608-62-9

IUPAC Name: 2-ethyloxirane
Synonyms: 1,2-butyleneoxide
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: RBACIKXCRWGCBB-UHFFFAOYNA-N
SMILES: CCC1CO1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)