Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-Butylene Oxide 98.0+%, TCI America™
SDP

Catalog No. B38335ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38335ML 5 mL
B383325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38335ML Supplier TCI America Supplier No. B38335ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30608-62-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD02683445
InChI Key RBACIKXCRWGCBB-BYPYZUCNSA-N
Synonym s---1,2-epoxybutane, 2s-2-ethyloxirane, s-1,2-epoxybutane, oxirane, ethyl-, 2s, s-ethyloxirane, 2beta-ethyloxirane, pubchem14100, s---ethyloxirane, s-1,2-epoxy-butane, s---butylene oxide
PubChem CID 10898629
IUPAC Name (2S)-2-ethyloxirane
SMILES CCC1CO1

Specifications

Color Colorless
Boiling Point 63°C
Quantity 5 mL
UN Number 3022
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-Butylene Oxide
EINECSNumber (2)-0229
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.