accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3061-65-2

H3COOCH2N

CAS RN 3061-65-2

IUPAC Name: 1-cyanoeth-1-en-1-yl acetate
Synonyms: 1-cyanoeth-1-en-1-yl acetate
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: LFSHREXVLSTLFB-UHFFFAOYSA-N
SMILES: CC(=O)OC(=C)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results