Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyanovinyl Acetate (stabilized with TBC) 97.0+%, TCI America™
SDP

Catalog No. A14775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14775G 5 g
A147725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14775G Supplier TCI America Supplier No. A14775G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3061-65-2
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00001868
InChI Key LFSHREXVLSTLFB-UHFFFAOYSA-N
Synonym 2-Acetoxyacrylonitrile, 1-Acetoxy-1-cyanoethylene, Acetic Acid 1-Cyanovinyl Ester
PubChem CID 18291
IUPAC Name 1-cyanoethenyl acetate
SMILES CC(=O)OC(=C)C#N

Specifications

Color Yellow
Boiling Point 173°C
Quantity 5 g
UN Number 3276
Formula Weight 111.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Cyanovinyl Acetate (stabilized with TBC)
RTECSNumber AG4550000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.