CAS RN 306279-65-2
IUPAC Name:
N-(2-ethoxyphenyl)-2-methoxybenzamide
Synonyms:
N-(2-ethoxyphenyl)-2-methoxybenzamide
Molecular Weight (g/mol):
271.32
Molecular Formula:
C16H17NO3
InChi Key:
IFOHCGGZEGVBME-UHFFFAOYSA-N
SMILES:
CCOC1=CC=CC=C1NC(=O)C1=CC=CC=C1OC
1
–
1
of
1
results
1
N-(2-Ethoxyphenyl)-2-methoxybenzamide, 97%, Thermo Scientific™
CAS: 306279-65-2 Molecular Formula: C16H17NO3 Molecular Weight (g/mol): 271.316 MDL Number: MFCD01010708 InChI Key: IFOHCGGZEGVBME-UHFFFAOYSA-N Synonym: n-2-ethoxyphenyl-2-methoxybenzamide,n-2-ethoxyphenyl 2-methoxyphenyl carboxamide PubChem CID: 700574 IUPAC Name: N-(2-ethoxyphenyl)-2-methoxybenzamide SMILES: CCOC1=CC=CC=C1NC(=O)C2=CC=CC=C2OC