accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOOCH3(E)

CAS RN 30633-94-4

IUPAC Name: (6E)-6-{[(4-butylphenyl)amino]methylidene}-3-methoxycyclohexa-2,4-dien-1-one
Synonyms: (6E)-6-{[(4-butylphenyl)amino]methylidene}-3-methoxycyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 283.37
Molecular Formula: C18H21NO2
InChi Key: NUBUXRIZVPWYJB-FYWRMAATSA-N
SMILES: CCCCC1=CC=C(N\C=C2/C=CC(OC)=CC2=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)