Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Methoxy-2-hydroxybenzylidene)-4-butylaniline 98.0+%, TCI America™
SDP

Catalog No. B02501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B02501G Supplier TCI America Supplier No. B02501G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30633-94-4
Molecular Formula C18H21NO2
Molecular Weight (g/mol) 283.371
MDL Number MFCD00059253
InChI Key NUBUXRIZVPWYJB-UHFFFAOYSA-N
Synonym N-(2-Hydroxy-p-anisal)-4-butylaniline, N-(2-Hydroxy-p-anisylidene)-4-butylaniline
PubChem CID 60150216
IUPAC Name 6-[(4-butylanilino)methylidene]-3-methoxycyclohexa-2,4-dien-1-one
SMILES CCCCC1=CC=C(C=C1)NC=C2C=CC(=CC2=O)OC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 283.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Methoxy-2-hydroxybenzylidene)-4-butylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.