accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHNCH3OOHO

CAS RN 3067-19-4

IUPAC Name: 2-acetamido-3-methylbutanoic acid
Synonyms: L-valine, N-acetyl-
Molecular Weight (g/mol): 159.19
Molecular Formula: C7H13NO3
InChi Key: IHYJTAOFMMMOPX-UHFFFAOYSA-N
SMILES: CC(C)C(NC(C)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results