Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-DL-valine 98.0+%, TCI America™
SDP

Catalog No. A012525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A012525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A012525G Supplier TCI America Supplier No. A012525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3067-19-4
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00066065
InChI Key IHYJTAOFMMMOPX-UHFFFAOYSA-N
Synonym n-acetyl-dl-valine, ac-dl-val-oh, n-acetylvaline, dl-valine, n-acetyl, 2-acetamido-3-methyl-butanoic acid, acetylvaline, n-acetylvaline #, valine, n-acetyl-, l, n-acetyl-valine, n-acetyl-d,l-valine
PubChem CID 227752
IUPAC Name 2-acetamido-3-methylbutanoic acid
SMILES CC(C)C(NC(C)=O)C(O)=O

Specifications

Melting Point 148°C
Color White
Quantity 25 g
Formula Weight 159.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-DL-valine
EINECSNumber (2)-1224
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.