accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 3068-88-0

IUPAC Name: 4-methyloxetan-2-one
Synonyms: 4-methyl-2-oxetanone
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: GSCLMSFRWBPUSK-UHFFFAOYNA-N
SMILES: CC1CC(=O)O1
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)