Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Butyrolactone, TCI America™
SDP

Catalog No. B0901500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0901500ML 500 mL
B090125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0901500ML Supplier TCI America Supplier No. B0901500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3068-88-0
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00005170
InChI Key GSCLMSFRWBPUSK-UHFFFAOYSA-N
Synonym 4-Methyl-2-oxetanone, beta-Methylpropiolactone
PubChem CID 18303
ChEBI CHEBI:82296
IUPAC Name 4-methyloxetan-2-one
SMILES CC1CC(=O)O1

Specifications

Melting Point -44°C
Color Yellow
Boiling Point 87°C
Quantity 500 mL
UN Number 1993
Formula Weight 86.09
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Butyrolactone
EINECSNumber (5)-0012
RTECSNumber RQ8050000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.