accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSNH2

CAS RN 306936-08-3

IUPAC Name: 2,3-dihydro-1-benzofuran-5-carbothioamide
Synonyms: 2,3-dihydro-1-benzofuran-5-carbothioamide
Molecular Weight (g/mol): 179.24
Molecular Formula: C9H9NOS
InChi Key: SOAROQIQNPHLJX-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC2=C(OCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results