accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 306936-12-9

IUPAC Name: ethyl 3-(chlorosulfonyl)pyridine-4-carboxylate
Synonyms: ethyl 3-(chlorosulfonyl)pyridine-4-carboxylate
Molecular Weight (g/mol): 249.67
Molecular Formula: C8H8ClNO4S
InChi Key: USJWOJZNWZSBGK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C=NC=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results