Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-(chlorosulfonyl)isonicotinate, 95%, Thermo Scientific™

Catalog No. SP00265CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SP00265CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SP00265CB Supplier Thermo Scientific Chemicals Supplier No. SP00265CB
Add to Cart

Chemical Identifiers

CAS 306936-12-9
Molecular Formula C8H8ClNO4S
Molecular Weight (g/mol) 249.665
InChI Key USJWOJZNWZSBGK-UHFFFAOYSA-N
Synonym ethyl 3-chlorosulfonyl isonicotinate, ethyl 3-chlorosulfonyl pyridine-4-carboxylate, ethyl-3-chlorosulphonylpyridine-4-carboxylate, ethyl 3-chlorosulphonyl pyridine-4-carboxylate, 4-pyridinecarboxylic acid, 3-chlorosulfonyl-, ethyl ester, 4-pyridinecarboxylic acid, 3-chlorosulfonyl-, ethyl ester 9ci
PubChem CID 2736400
IUPAC Name ethyl 3-chlorosulfonylpyridine-4-carboxylate
SMILES CCOC(=O)C1=C(C=NC=C1)S(=O)(=O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 249.67
Percent Purity 95%
Chemical Name or Material Ethyl 3-(chlorosulfonyl)isonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.