accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFFFFFFFFFFFFFFFF

CAS RN 307-59-5

IUPAC Name: hexacosafluorododecane
Synonyms: hexacosafluorododecane
Molecular Weight (g/mol): 638.09
Molecular Formula: C12F26
InChi Key: WNZGTRLARPEMIG-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results