accessibility menu, dialog, popup

UserName

Viewing profile as user:
IH3CNSSNH3C(Z)(E)(E/Z)(E/Z)

CAS RN 3071-70-3

IUPAC Name: 3-ethyl-2-[(5E)-7-[(2Z)-3-ethyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]hepta-1,3,5-trien-1-yl]-1,3-benzothiazol-3-ium iodide
Synonyms: 3-ethyl-2-[(5E)-7-[(2Z)-3-ethyl-1,3-benzothiazol-2-ylidene]hepta-1,3,5-trien-1-yl]-1,3-benzothiazol-3-ium iodide
Molecular Weight (g/mol): 544.51
Molecular Formula: C25H25IN2S2
InChi Key: OYVFJKVYVDYPFV-UHFFFAOYSA-M
SMILES: [I-].CCN1\C(SC2=CC=CC=C12)=C\C=C\C=CC=CC1=[N+](CC)C2=CC=CC=C2S1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)