accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(R)NHH3C

CAS RN 30727-18-5

IUPAC Name: (2R)-2-(ethoxycarbonyl)-1-methylpiperidin-1-ium
Synonyms: (2R)-2-(ethoxycarbonyl)-1-methylpiperidin-1-ium
Molecular Weight (g/mol): 172.25
Molecular Formula: C9H18NO2
InChi Key: ZICBNHLULHJETL-MRVPVSSYSA-O
SMILES: CCOC(=O)[C@H]1CCCC[NH+]1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results