Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-Methylpipecolate 97.0+%, TCI America™
SDP

Catalog No. E086425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E086425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E086425G Supplier TCI America Supplier No. E086425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30727-18-5
Molecular Formula C9H18NO2
Molecular Weight (g/mol) 172.25
MDL Number MFCD00006492
InChI Key ZICBNHLULHJETL-MRVPVSSYSA-O
Synonym ethyl 1-methylpipecolinate, ethyl 1-methyl-2-piperidinecarboxylate, ethyl n-methylpipecolinate, ethyl 1-methylpipecolate, 2-piperidinecarboxylic acid, 1-methyl-, ethyl ester, 1-methylpipecolic acid ethyl ester, pubchem11770, methyl 1-methyl piperidine-2-carboxylate, acmc-209hid, ksc495g6f
PubChem CID 566742
IUPAC Name (2R)-2-(ethoxycarbonyl)-1-methylpiperidin-1-ium
SMILES CCOC(=O)[C@H]1CCCC[NH+]1C

Specifications

Color Yellow
Boiling Point 95°C
Quantity 25 g
Formula Weight 171.24
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl 1-Methylpipecolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.