accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSNH2

CAS RN 307343-50-6

IUPAC Name: ethyl 2-amino-4-{[1,1'-biphenyl]-4-yl}thiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-{[1,1'-biphenyl]-4-yl}thiophene-3-carboxylate
Molecular Weight (g/mol): 323.41
Molecular Formula: C19H17NO2S
InChi Key: GGRXYEXAHRYAIO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results