accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOH3CCH3OOOHAl

CAS RN 30745-55-2

IUPAC Name: aluminium(3+) bis(2-ethylhexanoate) hydroxide
Synonyms: aluminium(3+) bis(2-ethylhexanoate) hydroxide
Molecular Weight (g/mol): 330.4
Molecular Formula: C16H31AlO5
InChi Key: ITXFWNWNWXSDBB-UHFFFAOYNA-K
SMILES: [OH-].[Al+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results