Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexanoato)hydroxyaluminum 96.0+%, TCI America™
SDP

Catalog No. B189525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B189525G 25 g
B1895250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B189525G Supplier TCI America Supplier No. B189525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30745-55-2
Molecular Formula C16H31AlO5
Molecular Weight (g/mol) 330.40
MDL Number MFCD00041940
InChI Key ITXFWNWNWXSDBB-UHFFFAOYNA-K
Synonym Hydroxyaluminum Bis(2-ethylhexanoate)
IUPAC Name aluminum(3+) bis(2-ethylhexanoate) hydroxide
SMILES [OH-].[Al+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O

Specifications

Color White
Quantity 25 g
UN Number 1325
Formula Weight 330.40
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-ethylhexanoato)hydroxyaluminum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.