accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2FFZnBr

CAS RN 307496-33-9

IUPAC Name: zinc(2+) (2,6-difluorophenyl)methanide bromide
Synonyms: zinc(2+) (2,6-difluorophenyl)methanide bromide
Molecular Weight (g/mol): 272.4
Molecular Formula: C7H5BrF2Zn
InChi Key: GCXVBPCJIZAPER-UHFFFAOYSA-M
SMILES: [Zn++].[Br-].[CH2-]C1=C(F)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results