accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHSOOS(E/Z)

CAS RN 307543-71-1

IUPAC Name: N-[(2-oxo-1,2-dihydronaphthalen-1-ylidene)methyl]thiophene-2-sulfonamide
Synonyms: N-[(2-oxonaphthalen-1-ylidene)methyl]thiophene-2-sulfonamide
Molecular Weight (g/mol): 317.38
Molecular Formula: C15H11NO3S2
InChi Key: AUXKGDNZANYAPX-UHFFFAOYSA-N
SMILES: O=C1C=CC2=CC=CC=C2C1=CNS(=O)(=O)C1=CC=CS1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results