accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3077-13-2

IUPAC Name: 1-[(2-hydroxypropyl)(phenyl)amino]propan-2-ol
Synonyms: 1-[(2-hydroxypropyl)(phenyl)amino]propan-2-ol
Molecular Weight (g/mol): 209.29
Molecular Formula: C12H19NO2
InChi Key: FKOMNQCOHKHUCP-UHFFFAOYNA-N
SMILES: CC(O)CN(CC(C)O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results