Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(2-hydroxypropyl)aniline (DL- and meso- mixture) 85.0+%, TCI America™
SDP

Catalog No. P079125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P079125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P079125G Supplier TCI America Supplier No. P079125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3077-13-2
Molecular Formula C12H19NO2
Molecular Weight (g/mol) 209.289
MDL Number MFCD00059141
InChI Key FKOMNQCOHKHUCP-UHFFFAOYSA-N
Synonym Anilinodi-2-propanol, 1,1′C-(Phenylimino)di-2-propanol
PubChem CID 76498
IUPAC Name 1-[N-(2-hydroxypropyl)anilino]propan-2-ol
SMILES CC(CN(CC(C)O)C1=CC=CC=C1)O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 209.29
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Bis(2-hydroxypropyl)aniline (DL- and meso- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.