accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NO

CAS RN 3077-16-5

IUPAC Name: 4-(4-methylphenyl)morpholine
Synonyms: 4-(4-methylphenyl)morpholine
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: OLAFVASCPJETBP-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N1CCOCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results