Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(p-Tolyl)morpholine 97.0+%, TCI America™
SDP

Catalog No. T22155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22155G Supplier TCI America Supplier No. T22155G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3077-16-5
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
InChI Key OLAFVASCPJETBP-UHFFFAOYSA-N
Synonym 4-Morpholinotoluene
PubChem CID 766255
IUPAC Name 4-(4-methylphenyl)morpholine
SMILES CC1=CC=C(C=C1)N2CCOCC2

Specifications

Melting Point 50°C
Color Yellow
Quantity 5 g
Formula Weight 177.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(p-Tolyl)morpholine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.