accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3078-09-9

IUPAC Name: (2E)-1-(4-nitrophenyl)-2-(phenylmethylidene)hydrazine
Synonyms: (2E)-1-(4-nitrophenyl)-2-(phenylmethylidene)hydrazine
Molecular Weight (g/mol): 241.25
Molecular Formula: C13H11N3O2
InChi Key: NOIFWEYOLLHIMW-GXDHUFHOSA-N
SMILES: [O-][N+](=O)C1=CC=C(N\N=C\C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results