Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzaldehyde 4-Nitrophenylhydrazone 98.0+%, TCI America™
SDP

Catalog No. B000925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B000925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B000925G Supplier TCI America Supplier No. B000925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3078-09-9
Molecular Formula C13H11N3O2
Molecular Weight (g/mol) 241.25
MDL Number MFCD00024596
InChI Key NOIFWEYOLLHIMW-GXDHUFHOSA-N
Synonym Benzal-4-nitrophenylhydrazone
PubChem CID 6436771
IUPAC Name N-[(E)-benzylideneamino]-4-nitroaniline
SMILES C1=CC=C(C=C1)C=NNC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 195°C
Quantity 25 g
Formula Weight 241.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Benzaldehyde 4-Nitrophenylhydrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.