accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONCO

CAS RN 30806-83-8

IUPAC Name: ethyl 4-isocyanatobenzoate
Synonyms: ethyl 4-isocyanatobenzoate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: CFEPCPHKICBCJV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)N=C=O
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results