CAS RN 308080-99-1
Molecular sieves 3A, 8 to 12 mesh
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular sieves, 3A, powder
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular sieves, 3A, 1-2mm (0.04-0.08in) dia. pellets
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular sieves 3A, powder <50 micron
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular sieves 3A, 4 to 8 mesh
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular sieves, 3A, 3-4mm (0.12-0.16in) dia. pellets
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular sieves, Solstice
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular Sieves, 3 Å, 4 to 8mesh, Solstice
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular Sieves, 3 Å, 8 to 12mesh, Solstice
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular Sieves 3A, Pellets, Dia.: 3.2mm, Solstice
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Molecular Sieves, 3A, Pellets, Dia.: 1.6mm, Solstice
CAS: 308080-99-1 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 MDL Number: MFCD00131613 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]