accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH

CAS RN 308081-08-5

IUPAC Name: 5-(2-ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole
Synonyms: atipamezole atipamezol atipamezolum 阿替美唑 antisedan
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: HSWPZIDYAHLZDD-UHFFFAOYSA-N
SMILES: CCC1(CC2=CC=CC=C2C1)C1=CN=CN1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results