accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30825-88-8

IUPAC Name: methyl 5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Synonyms: methyl 5-bromo-2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate
Molecular Weight (g/mol): 249.02
Molecular Formula: C6H5BrN2O4
InChi Key: UJCPEPQYSPRWGU-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Br)C(=O)NC(=O)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results