accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)O(R)(S)OH(S)HONONHO

CAS RN 3083-77-0

IUPAC Name: 1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: arauridine
Molecular Weight (g/mol): 244.2
Molecular Formula: C9H12N2O6
InChi Key: DRTQHJPVMGBUCF-CCXZUQQUSA-N
SMILES: OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)