accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3084-47-7

IUPAC Name: 1-(dipentylphosphoryl)pentane
Synonyms: 1-(dipentylphosphoryl)pentane
Molecular Weight (g/mol): 260.4
Molecular Formula: C15H33OP
InChi Key: SAIZTOCXWYRRNW-UHFFFAOYSA-N
SMILES: CCCCCP(=O)(CCCCC)CCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results