Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-amylphosphine Oxide, MP Biomedicals

Catalog No. ICN227284 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227284 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227284 Supplier MP Biomedicals, Inc Supplier No. 0522728401

Chemical Identifiers

CAS 3084-47-7
Molecular Formula C15H33OP
Molecular Weight (g/mol) 260.402
InChI Key SAIZTOCXWYRRNW-UHFFFAOYSA-N
Synonym tripentylphosphine oxide, phosphine oxide, tripentyl, tri-n-amyl phosphine oxide, diamyl amyl phosphine oxide, 1-dipentylphosphoryl pentane, wln: op5&5&5
PubChem CID 99394
IUPAC Name 1-dipentylphosphorylpentane
SMILES CCCCCP(=O)(CCCCC)CCCCC

Specifications

Quantity 1 g
Chemical Name or Material Tri-n-Amyl Phosphine Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.