CAS RN 30860-22-1
IUPAC Name:
3-chlorobicyclo[2.2.1]heptan-2-one
Synonyms:
3-chlorobicyclo[2.2.1]heptan-2-one
Molecular Weight (g/mol):
144.6
Molecular Formula:
C7H9ClO
InChi Key:
PQRKEKMZLKKQOP-UHFFFAOYNA-N
SMILES:
ClC1C2CCC(C2)C1=O
1
–
1
of
1
results
1
3-Chloro-2-norbornanone 97%, Thermo Scientific™
CAS: 30860-22-1 Molecular Formula: C7H9ClO Molecular Weight (g/mol): 144.598 InChI Key: PQRKEKMZLKKQOP-UHFFFAOYSA-N Synonym: 3-chloro-2-norbornanone,2-norbornanone, 3-chloro,2-chlorobicyclo 2.2.1 heptan-3-one,3-chlorobicyclo 2.2.1 heptan-2-one,bicyclo 2.2.1 heptan-2-one, 3-chloro-, endo,3-chloronorbornan-2-one,bicyclo 2.2.1 heptan-2-one, 3-chloro,acmc-1ad3y,pqrkekmzlkkqop-uhfffaoysa PubChem CID: 98217 IUPAC Name: 2-chlorobicyclo[2.2.1]heptan-3-one SMILES: C1CC2CC1C(C2=O)Cl