Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-norbornanone 97%, Thermo Scientific™

Catalog No. AC109760050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109760050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC109760050 Supplier Thermo Scientific Chemicals Supplier No. 109760050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30860-22-1
Molecular Formula C7H9ClO
Molecular Weight (g/mol) 144.598
InChI Key PQRKEKMZLKKQOP-UHFFFAOYSA-N
Synonym 3-chloro-2-norbornanone, 2-norbornanone, 3-chloro, 2-chlorobicyclo 2.2.1 heptan-3-one, 3-chlorobicyclo 2.2.1 heptan-2-one, bicyclo 2.2.1 heptan-2-one, 3-chloro-, endo, 3-chloronorbornan-2-one, bicyclo 2.2.1 heptan-2-one, 3-chloro, acmc-1ad3y, pqrkekmzlkkqop-uhfffaoysa
PubChem CID 98217
IUPAC Name 2-chlorobicyclo[2.2.1]heptan-3-one
SMILES C1CC2CC1C(C2=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Infrared Spectrum Authentic
Refractive Index 1.4980 to 1.5000 (20°C, 589nm)
Quantity 5 g
Formula Weight 144.6
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-norbornanone, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.