accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOCH3HOCH3HOCH3HOTi

CAS RN 3087-37-4

IUPAC Name: tetrakis(propan-1-ol) titanium
Synonyms: tetrakis(propanol) titanium
Molecular Weight (g/mol): 288.25
Molecular Formula: C12H32O4Ti
InChi Key: XTTBFCWRLDKOQU-UHFFFAOYSA-N
SMILES: [Ti].CCCO.CCCO.CCCO.CCCO
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)