CAS RN 3088-41-3
IUPAC Name:
1-(2-cyanoethyl)piperidin-1-ium
Synonyms:
1-(2-cyanoethyl)piperidin-1-ium
Molecular Weight (g/mol):
139.22
Molecular Formula:
C8H15N2
InChi Key:
YZICFVIUVMCCOC-UHFFFAOYSA-O
SMILES:
N#CCC[NH+]1CCCCC1
1
–
1
of
1
results
1
1-(2-Cyanoethyl)piperidine 98.0+%, TCI America™
CAS: 3088-41-3 Molecular Formula: C8H14N2 Molecular Weight (g/mol): 138.214 MDL Number: MFCD00006511 InChI Key: YZICFVIUVMCCOC-UHFFFAOYSA-N Synonym: 1-piperidinepropanenitrile,1-piperidinepropionitrile,3-piperidin-1-yl propanenitrile,1-2-cyanoethyl piperidine,3-piperidinopropionitrile,beta-piperidinopropionitrile,3-1-piperidine propionitrile,3-piperidin-1-yl propionitrile,3-n-piperidino propionitrile,3-piperidylpropanenitrile PubChem CID: 18338 IUPAC Name: 3-piperidin-1-ylpropanenitrile SMILES: C1CCN(CC1)CCC#N