Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Cyanoethyl)piperidine 98.0+%, TCI America™
SDP

Catalog No. C131325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C131325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C131325G Supplier TCI America Supplier No. C131325G
 Restricted Item
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3088-41-3
Molecular Formula C8H14N2
Molecular Weight (g/mol) 138.214
MDL Number MFCD00006511
InChI Key YZICFVIUVMCCOC-UHFFFAOYSA-N
Synonym 1-piperidinepropanenitrile, 1-piperidinepropionitrile, 3-piperidin-1-yl propanenitrile, 1-2-cyanoethyl piperidine, 3-piperidinopropionitrile, beta-piperidinopropionitrile, 3-1-piperidine propionitrile, 3-piperidin-1-yl propionitrile, 3-n-piperidino propionitrile, 3-piperidylpropanenitrile
PubChem CID 18338
IUPAC Name 3-piperidin-1-ylpropanenitrile
SMILES C1CCN(CC1)CCC#N

Specifications

Boiling Point 105°C
Quantity 25 g
UN Number 3276
Formula Weight 138.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Cyanoethyl)piperidine
RTECSNumber TN3714000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.