accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClCl

CAS RN 309-00-2

IUPAC Name: 1,8,9,10,11,11-hexachlorotetracyclo[6.2.1.1³,⁶.0²,⁷]dodeca-4,9-diene
Synonyms: aldrin (iso)
Molecular Weight (g/mol): 364.9
Molecular Formula: C12H8Cl6
InChi Key: QBYJBZPUGVGKQQ-UHFFFAOYNA-N
SMILES: ClC1=C(Cl)C2(Cl)C3C4CC(C=C4)C3C1(Cl)C2(Cl)Cl
Grade 
  • (2)
Element 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)